Learn More
Kuhner Shaker Inc 96 WELL ROUND LOW GLUCOSE FEED
Catalog No. NC2209797
Encompass
FeedPlate, Glucose, 96 Well, Round, 3 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
This item is not returnable. View return policy
Catalog No. NC2209797 Supplier Kuhner Shaker Inc Supplier No. SMFP00701
Add to Cart
Add to Cart
FeedPlate, Glucose, 96 Well, Round, 3 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!