Learn More
Alkali Scientific 96 Well Round Silicone Sealing Mat, Autoclavable, 10/pk, 50/cs
SDP
Catalog No. 502388926
Encompass_Preferred
Round capped mats for sealing 96-well storage plates. Alphanumeric grid-referencing. Molded from a proprietary thermoplastic elastomer for a flexible design. Suitable for long term storage. Available in autoclavable format. Solvent resistant (including DMSO, Ethanol, and Isopropanol). Suitable for UV Irradiation and Gamma Irradiation. Temperature Range: −20°C to +55°C.
Catalog No. 50-238-8926 Supplier Alkali Scientific Supplier No. MPD51
Round capped mats for sealing 96-well storage plates. Alphanumeric grid-referencing. Molded from a proprietary thermoplastic elastomer for a flexible design. Suitable for long term storage. Available in autoclavable format. Solvent resistant (including DMSO, Ethanol, and Isopropanol). Suitable for UV Irradiation and Gamma Irradiation. Temperature Range: −20°C to +55°C.