Learn More
Waters Corp Waters 96-well Sample Collection Plate, 2 mL Square Well, Polypropylene, SBS/ANSI Compliant
Catalog No. 50781141 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well sample collection plate, made from polypropylene with 2 mL square wells, is SBS/ANSI compliant for robot systems. It can function as a collection plate for SPE and filtration formats, is suitable for autosamplers, and withstands up to 5000g centrifugal force. With a sealant, it protects samples from contamination, temperature changes, and organic solvents.
  • SBS/ANSI compliant for robot systems.
  • Usable as a collection plate for SPE and filtration formats.
  • Designed for autosamplers.
  • Manufactured from polypropylene.
  • Features square wells.
  • Maximum centrifugal force of 5000g.
  • Protects samples from contamination, temperature fluctuations, and organic solvents when used with a sealant.
  • DNase- and RNase-free.
Catalog No. 50-781-141 Supplier Waters Corp Supplier No. 186002482
This 96-well sample collection plate, made from polypropylene with 2 mL square wells, is SBS/ANSI compliant for robot systems. It can function as a collection plate for SPE and filtration formats, is suitable for autosamplers, and withstands up to 5000g centrifugal force. With a sealant, it protects samples from contamination, temperature changes, and organic solvents.
  • SBS/ANSI compliant for robot systems.
  • Usable as a collection plate for SPE and filtration formats.
  • Designed for autosamplers.
  • Manufactured from polypropylene.
  • Features square wells.
  • Maximum centrifugal force of 5000g.
  • Protects samples from contamination, temperature fluctuations, and organic solvents when used with a sealant.
  • DNase- and RNase-free.