Learn More
Waters Corp Waters Sample Collection Plate, 96-well, 700 μL, Round Well, Polypropylene
Catalog No. 50201497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well Sample Collection Plate is designed for laboratory applications. It features 700 μL round wells and is made from polypropylene, ensuring chemical inertness.
  • SBS/ANSI compliant for robot compatible systems
  • Can serve as a collection plate for 96-well SPE and filtration-plate formats
  • Manufactured using polypropylene to ensure no chemical reaction when storing analytes
  • Round wells
  • Can be centrifuged to a maximum centrifugal force of 5000g
  • Protects samples and analytes from contamination and temperature fluctuation when used with a sealant
  • Sealant protects from organic solvents
  • Critical component in ACQUITY UPLC System for improved resolution, sensitivity, and throughput
  • Polypropylene has a high melting point and does not react with water, detergents, acids, or bases, making it resistant to disintegration
Catalog No. 50-201-497 Supplier Waters Corp Supplier No. 186005837
This 96-well Sample Collection Plate is designed for laboratory applications. It features 700 μL round wells and is made from polypropylene, ensuring chemical inertness.
  • SBS/ANSI compliant for robot compatible systems
  • Can serve as a collection plate for 96-well SPE and filtration-plate formats
  • Manufactured using polypropylene to ensure no chemical reaction when storing analytes
  • Round wells
  • Can be centrifuged to a maximum centrifugal force of 5000g
  • Protects samples and analytes from contamination and temperature fluctuation when used with a sealant
  • Sealant protects from organic solvents
  • Critical component in ACQUITY UPLC System for improved resolution, sensitivity, and throughput
  • Polypropylene has a high melting point and does not react with water, detergents, acids, or bases, making it resistant to disintegration