Learn More
Waters Corp WATERS CORPORATION Sample Collection Plate, 96-Well, 800 μL, Round Well
Catalog No. 50781140 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well sample collection plate features 800 μL round wells. It is SBS/ANSI compliant for robot-compatible systems and functions as a collection plate for 96-well SPE and filtration-plate formats.
  • SBS/ANSI compliant for robot compatible systems
  • Functions as a collection plate for 96-well SPE and filtration plate formats
  • Designed for high-throughput applications
  • Compatible with most automated liquid handling systems
  • Suitable for both sample storage and analysis
  • High-quality polypropylene ensures excellent chemical resistance and sample integrity
Catalog No. 50-781-140 Supplier Waters Corp Supplier No. 186002481
This 96-well sample collection plate features 800 μL round wells. It is SBS/ANSI compliant for robot-compatible systems and functions as a collection plate for 96-well SPE and filtration-plate formats.
  • SBS/ANSI compliant for robot compatible systems
  • Functions as a collection plate for 96-well SPE and filtration plate formats
  • Designed for high-throughput applications
  • Compatible with most automated liquid handling systems
  • Suitable for both sample storage and analysis
  • High-quality polypropylene ensures excellent chemical resistance and sample integrity