Learn More
Thermo Scientific™ 96-Well Semi-Skirted PCR Plates
Catalog No. 21402454 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Natural
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-454 White
21-402-375 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-454 Supplier Thermo Scientific™ Supplier No. 3484W
Break plates apart into 24- or 48-well formats with the special design of these 96-well semi-skirted PCR plates, made from 100% pure polypropylene.
Break plates apart into 24- or 48-well formats with the special design of these Thermo Scientific™ 96-well semi-skirted PCR plates, made from 100% pure polypropylene.
  • Thin-wall design for optimal heat transfer
  • Elevated rims for secure sealing
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color White
Product Type PCR Microplate
Skirt Style Semi
Material Polypropylene
No. of Wells 96