Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 96-Well Semi-Skirted PCR Plates

Catalog No. p-4525312
Change view
Click to view available options
Color:
Natural
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-375 Natural
21-402-454 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-375 Supplier Thermo Scientific™ Supplier No. 3484

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Break plates apart into 24- or 48-well formats with the special design of these 96-well semi-skirted PCR plates, made from 100% pure polypropylene.

Break plates apart into 24- or 48-well formats with the special design of these Thermo Scientific™ 96-well semi-skirted PCR plates, made from 100% pure polypropylene.

  • Thin-wall design for optimal heat transfer
  • Elevated rims for secure sealing
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Packaging 50 Pk.
Color Natural
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Semi
Material Polypropylene
No. of Wells 96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.