Learn More
LUNA NANOTECH INC 96-Well Silica Membrane Plate, Full-Skirted
Catalog No. 502566914
Encompass_Preferred
- Speed up your RNA and DNA processing purification by going high throughput!
- Each well features a thick silica membrane to maximize nucleic acid binding and recovery of RNA and DNA from many sample types.
- Our Purification Plate allows the user to process up to 96 samples simultaneously.
- Processing can be done via centrifugation or vaccuum.
- Available in Full-skirted and Semi-skirted format.
Catalog No. 50-256-6914 Supplier LUNA NANOTECH INC Supplier No. USP096F
- Speed up your RNA and DNA processing purification by going high throughput!
- Each well features a thick silica membrane to maximize nucleic acid binding and recovery of RNA and DNA from many sample types.
- Our Purification Plate allows the user to process up to 96 samples simultaneously.
- Processing can be done via centrifugation or vaccuum.
- Available in Full-skirted and Semi-skirted format.