Learn More
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Ideally suited for agglutination tests
- No sharp corners, which means that they can be pipetted easily and cleanly
- Suitable for +/- analyses
- Solid bottom
Specifications
Specifications
| Barcode | Alphanumeric |
| Certified | Free of human DNA and pyrogens |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Product Type | Assay Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
