Learn More
Greiner Bio-One 96-well Solid Bottomed Microplates
GREENER_CHOICE
Catalog No. p-7075498 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-000-125 Flat Bottom (F-well)
07-000-132 Round Bottom (U-well)
07-000-142 Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-125 Supplier Greiner Bio-One Supplier No. 655161
Add to Cart
Edge
Add to Cart
For high-throughput screening and assay
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Ideally suited for agglutination tests
  • No sharp corners, which means that they can be pipetted easily and cleanly
  • Suitable for +/- analyses
  • Solid bottom
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Certified Free of human DNA and pyrogens
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Product Type Assay Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 0.025 to 0.340 mL
Sterility Sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 2
Quantity 100/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 14 mm