Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ Splash-Free 96-Well Base

Catalog No. 4312063
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Bases
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-120-63 10 Bases
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-120-63 Supplier Applied Biosystems™ Supplier No. 4312063
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 MicroAmp Splash Free 96-Well Bases.

Used for centrifugation

  • Designed to hold MicroAmp reaction tubes in MicroAmp tray/retainer sets or 96-well reaction plates for convenient pre-PCR and post-PCR sample processing
  • Uniform Results
  • Use with instruments that have heated sample covers to eliminate condensation and need for mineral oil overlay
  • Proprietary MicroAmp reaction tubes are essential for high-performance PCR to ensure maximum temperature and amplification uniformity for samples

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ Splash Free 96-Well Bases. Store at room temperature.
Product Line MicroAmp
Color Black
Product Type 96-Well Base
For Use With (Equipment) 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 10 Bases
Shipping Condition Room Temperature
Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.