Learn More
Arcticwhite LLC ArctiSeal 96 Well Square, White Silicone Sealing Mat, w/ PTFE Coating, 5 Plate Seals per Pack
SDP
Catalog No. NC9385013
Encompass
ArctiSeal 96 Well Square, White Silicone Sealing Mat, PTFE Coating, 5 Plate Seals per Pack. A silicone sealing mat with a PTFE coating for sealing 96-well square-well microplates. Format - 96-well. Well shape - square. Supplied by ArcticWhite as 5 plate seals per pack.
Catalog No. NC9385013 Supplier Arcticwhite LLC Supplier No. AWSM1003S
ArctiSeal 96 Well Square, White Silicone Sealing Mat, PTFE Coating, 5 Plate Seals per Pack. A silicone sealing mat with a PTFE coating for sealing 96-well square-well microplates. Format - 96-well. Well shape - square. Supplied by ArcticWhite as 5 plate seals per pack.