Learn More
Alkali Scientific ASI™ 96 Well Storage Plate, PP, Round Bottom, Clear, 100/Cs
SDP
Catalog No. 503027922
Encompass_Preferred
ASI™ 96 Well Storage Plates are designed for storage of biological samples and chemicals. Lids included Storage of biological samples and chemicals of small quantity Temperature range: -70 ~ +140 °C High chemically resistant for such as organic solvents, strong acids and strong bases
Catalog No. 50-302-7922 Supplier Alkali Scientific Supplier No. SPR0096
Add to Cart
Add to Cart
ASI™ 96 Well Storage Plates are designed for storage of biological samples and chemicals. Lids included Storage of biological samples and chemicals of small quantity Temperature range: -70 ~ +140 °C High chemically resistant for such as organic solvents, strong acids and strong bases