Learn More
Andwin Scientific 96-WELL TEMPPLATE II PP PCR P
SDP
Catalog No. 50819656
Encompass_Preferred
The 96-well PCR plate feature high chimney wells that help reduce the chance of cross-contamination. The wells can be sealed with either strip caps or mats. This product is made with polypropylene that permits a quick and even heat transfer for reliable amplifications. It is free of detectable RNase, DNase, DNA, pyrogen, and PCR inhibitors.
Catalog No. 50-819-656 Supplier Andwin Scientific Supplier No. 14029600
The 96-well PCR plate feature high chimney wells that help reduce the chance of cross-contamination. The wells can be sealed with either strip caps or mats. This product is made with polypropylene that permits a quick and even heat transfer for reliable amplifications. It is free of detectable RNase, DNase, DNA, pyrogen, and PCR inhibitors.