Learn More
Andwin Scientific 96-WELL TEMPPLATE V LOW PROF
SDP
Catalog No. 50819661
Encompass_Preferred
The 96-well PCR plate is a full-skirted low profile plate, which is a recommended feature for volumes of less than 50 l. It has easy-to-read black alphanumeric labels that permit users to easily identify samples. The wells are rimmed to reduce the chance of cross contamination. Its polypropylene material allows quick and even heat transfer for reliable amplifications.
Catalog No. 50-819-661 Supplier Andwin Scientific Supplier No. 14029800
The 96-well PCR plate is a full-skirted low profile plate, which is a recommended feature for volumes of less than 50 l. It has easy-to-read black alphanumeric labels that permit users to easily identify samples. The wells are rimmed to reduce the chance of cross contamination. Its polypropylene material allows quick and even heat transfer for reliable amplifications.