Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 96-well tips, 50 μL
GREENER_CHOICE

Catalog No. 14222375 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-375 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-375 Supplier Axygen™ Supplier No. RRP50CR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Axygen™ offers a comprehensive line of tips for PerkinElmer™ automation systems.

These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in non-sterile, sterile, non-filtered and filtered. Clear and Conductive Black polypropylene tips are available in 96-well format designed for PerkinElmer™ Janus™/MultiProbe Variospan-1, 4, 8 head.

  • 50 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • non-sterile
  • Compatibility: PerkinElmer™ Janus™/MultiProbe Variospan-1, 4, 8 head
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Standard, Round
Packaging SLAS Rack
Quantity 96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs
For Use With (Equipment) PerkinElmer, Tecan
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.