Learn More
CELLTREAT Scientific Products 96 Well Round Bottom Non-treated Plate, Sterile
SDP
Catalog No. 50202148
Encompass_Preferred
CELLTREAT Scientific Products 96 Well Non-Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Round bottom with working volume of 50-300¿L. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.
Catalog No. 50-202-148 Supplier CELLTREAT Scientific Products Supplier No. 229590
Add to Cart
Add to Cart
CELLTREAT Scientific Products 96 Well Non-Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Round bottom with working volume of 50-300¿L. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.