Learn More
Bellco Glass, Inc. 96 Well Tissue Culture Plate
SDP
Supplier:  Bellco Glass, Inc. 229197000
Encompass_Preferred
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack Description5/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.33cm²RNase/DNase FreeYes
Catalog No. 50-233-0144