Learn More
CELLTREAT Scientific Products 96 Well Tissue Culture Plate, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229196
Encompass_Preferred
CELLTREAT Scientific Products 96 Well Tissue Culture Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with cell growth area of 0.33cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.
Catalog No. 50-202-142