Learn More
Thomas Scientific 96 Well Tissue Culture Plate with Lid, Individual, Sterile
SDP
Catalog No. 501461452
Encompass
Available in 6, 12, 24, 48 and 96 well flat bottom configurations NEW — 96 well plates now available with a round bottom Plates are labeled with alpha-numeric markings for easy identification of individual wells Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination Lids have a stacking ring for added stability when handling tall stacks of plates All multiple well plates are packaged sterile (gamma irradiated) NEW – Now available in 4 well flat bottom, treated or non-treated configurations Thomas No. 1177Y90 and 1177Y96 NEW – Now available in 1 well configuration Useful for high volume and robotic screening of DNA libraries, as well as manual plating Same footprint as standard multiple well plates Suitable for dot-blotting bacterial clones on 11.4 x 7.3 cm (4.5 x 2.875 inches) nylon membranes Lids contain molded spacers to allow gas exchange and stacking rings for increased stab
This item is not returnable. View return policy
Catalog No. 50-146-1452 Supplier Thomas Scientific Supplier No. 1228K43
Available in 6, 12, 24, 48 and 96 well flat bottom configurations NEW — 96 well plates now available with a round bottom Plates are labeled with alpha-numeric markings for easy identification of individual wells Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination Lids have a stacking ring for added stability when handling tall stacks of plates All multiple well plates are packaged sterile (gamma irradiated) NEW – Now available in 4 well flat bottom, treated or non-treated configurations Thomas No. 1177Y90 and 1177Y96 NEW – Now available in 1 well configuration Useful for high volume and robotic screening of DNA libraries, as well as manual plating Same footprint as standard multiple well plates Suitable for dot-blotting bacterial clones on 11.4 x 7.3 cm (4.5 x 2.875 inches) nylon membranes Lids contain molded spacers to allow gas exchange and stacking rings for increased stab