Learn More
Molecular Devices Molecular Devices Cell Wash Head, 384-Well, Angled Dispense Pins, Quick-Change
Catalog No. 5000014738
Encompass_Preferred
Replacement 384-well cell wash head designed for use with Molecular Devices AquaMax 2000 and 4000 microplate washers. The wash head delivers wash solutions to the sides of wells using angled dispense pins to minimize cell loss and preserve assay performance in cell-based assays. It installs quickly and is interchangeable with other AquaMax wash heads for flexible operation.
  • Designed for 384-well microplates
  • Angled dispense pins to minimize cell disturbance
  • High cell retention with optimized wash settings
  • Tool-free, quick interchange with other wash heads
  • Optimized for gentle washing in cell-based assays
Catalog No. 50-000-14738 Supplier Molecular Devices Supplier No. 03105218T
Add to Cart
Add to Cart
Request a Quote
Replacement 384-well cell wash head designed for use with Molecular Devices AquaMax 2000 and 4000 microplate washers. The wash head delivers wash solutions to the sides of wells using angled dispense pins to minimize cell loss and preserve assay performance in cell-based assays. It installs quickly and is interchangeable with other AquaMax wash heads for flexible operation.
  • Designed for 384-well microplates
  • Angled dispense pins to minimize cell disturbance
  • High cell retention with optimized wash settings
  • Tool-free, quick interchange with other wash heads
  • Optimized for gentle washing in cell-based assays