Learn More
Alkali Scientific 96 Well White Plate, PS/PS, TC treated, Sterile 5/25
SDP
Catalog No. 502335932
Encompass_Preferred
White plate / clear bottom. Sterile. Ideal plates for obtaining amplified signals in a luminescence assays while minimizing cross-talk. The highly transparent bottom provides an optimal environment for microscopic observation. Optimized surface technology for cell attachment and microscopic observation. Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment. ▪ White Wall and topside (PS) / Clear Bottom ▪ Bottom material: Optically Clear PS ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ sterile 5/pack ▪ Non – pyrogenic ▪ Non – cytotoxic ▪ DNase / RNase – free ▪ Human DNA – free
Catalog No. 50-233-5932 Supplier Alkali Scientific Supplier No. TPW0096
Add to Cart
Add to Cart
White plate / clear bottom. Sterile. Ideal plates for obtaining amplified signals in a luminescence assays while minimizing cross-talk. The highly transparent bottom provides an optimal environment for microscopic observation. Optimized surface technology for cell attachment and microscopic observation. Tissue Culture Treated for cell adhesion and attachment providing an ideal proliferation environment. ▪ White Wall and topside (PS) / Clear Bottom ▪ Bottom material: Optically Clear PS ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ sterile 5/pack ▪ Non – pyrogenic ▪ Non – cytotoxic ▪ DNase / RNase – free ▪ Human DNA – free