Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotage 960 MICROLITER TIPS AND HOLDER
SDP

Catalog No. NC1236896
Encompass

  • Tip volume: 1000 µL
  • Quantity: 960 tips
  • Color: Clear
  • Compatibility: Biotage Extrahera LV-200, Biotage Extrahera Classic
  • Packaging: Racked (10 x 96 tips)
  • Application: Automated liquid handling, sample preparation
  • Product type: Pipette tips
  • Dispensing accuracy: Up to 1000 µL in one aliquot (system dependent)
  • Tip type: Disposable

Catalog No. NC1236896 Supplier Biotage Supplier No. 414141
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Tip volume: 1000 µL
  • Quantity: 960 tips
  • Color: Clear
  • Compatibility: Biotage Extrahera LV-200, Biotage Extrahera Classic
  • Packaging: Racked (10 x 96 tips)
  • Application: Automated liquid handling, sample preparation
  • Product type: Pipette tips
  • Dispensing accuracy: Up to 1000 µL in one aliquot (system dependent)
  • Tip type: Disposable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.