Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Black Rubber Vacuum, 3/8"" ID x 7/8"" OD; 25 ft
Catalog No. NC0633990
Encompass
The tubing is ideal for vacuum and pressure applications for general lab and industrial use. It features thick flexible walls with a temperature range of 0 to 150¿F (-18 to 65¿C). Can be sterilized with ethylene oxide or autoclave for 30 minutes at 15 psi (1.0 bar).
Catalog No. NC0633990 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9614309
The tubing is ideal for vacuum and pressure applications for general lab and industrial use. It features thick flexible walls with a temperature range of 0 to 150¿F (-18 to 65¿C). Can be sterilized with ethylene oxide or autoclave for 30 minutes at 15 psi (1.0 bar).