Learn More
Sklar Instruments Econo Sterile Iris Scissors, Sklar , 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. CS/50.
SDP
Catalog No. NC9684578 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Sterile Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. BX50.
Catalog No. NC9684578 Supplier Sklar Instruments Supplier No. 962506
Econo Sterile Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. BX50.