Learn More
Hach Company Sension+ 9650 Sodium Ion Selective Electrode (ISE) – sensor only
Catalog No. 501998017 Shop All Hach Company Products
Encompass
Sension+ 9650 is a half-cell (reference not integrated) sodium ion selective electrode (ISE) glass body and glass membrane selective to sodium ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9650 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9650 is ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-8017 Supplier Hach Company Supplier No. LZW9650.97.0002
Sension+ 9650 is a half-cell (reference not integrated) sodium ion selective electrode (ISE) glass body and glass membrane selective to sodium ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9650 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9650 is ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.