Learn More
Hach Company Sension+ 9652 Chloride Ion Selective Electrode (ISE) – sensor only
Catalog No. 501998018 Shop All Hach Company Products
Encompass
Sension+ 9652 is a half-cell (reference not integrated) chloride ion selective electrode (ISE) with an epoxy body and non-replaceable crystalline membrane selective to chloride ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9652 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9652 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-8018 Supplier Hach Company Supplier No. LZW9652.97.0002
Sension+ 9652 is a half-cell (reference not integrated) chloride ion selective electrode (ISE) with an epoxy body and non-replaceable crystalline membrane selective to chloride ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9652 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9652 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.