Learn More
Hach Company Sension+ 9655 Fluoride Ion Selective Electrode (ISE) – sensor only
Catalog No. 501998021 Shop All Hach Company Products
Encompass
Sension+ 9655 is a half-cell (reference not integrated) fluoride ion selective electrode (ISE) with an epoxy body and non-replaceable crystalline membrane selective to fluoride ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9655 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9655 is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality applications.
Catalog No. 50-199-8021 Supplier Hach Company Supplier No. LZW9655.97.0002
Sension+ 9655 is a half-cell (reference not integrated) fluoride ion selective electrode (ISE) with an epoxy body and non-replaceable crystalline membrane selective to fluoride ions in solution. It is recommended for use with the Sension+ 5044 reference electrode. The 9655 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 9655 is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality applications.