Learn More
Hach Company Sension+ 9655 Fluoride Combination Ion Selective Electrode (ISE)
Catalog No. 501998023 Shop All Hach Company Products
Encompass
Sension+ 9655C is a combination fluoride ion selective electrode (ISE) with an epoxy body, non-refillable, single-junction reference, and non-replaceable crystalline membrane selective to fluoride ions in solution. The 9655C has a BNC connector for laboratory use with the Hach Sension+ MM340 GLP pH/ISE Laboratory Meter or the Sension+ MM374 Laboratory Multimeter. The 9655C is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality laboratory applications.
Catalog No. 50-199-8023 Supplier Hach Company Supplier No. LZW9655C.97.002
Sension+ 9655C is a combination fluoride ion selective electrode (ISE) with an epoxy body, non-refillable, single-junction reference, and non-replaceable crystalline membrane selective to fluoride ions in solution. The 9655C has a BNC connector for laboratory use with the Hach Sension+ MM340 GLP pH/ISE Laboratory Meter or the Sension+ MM374 Laboratory Multimeter. The 9655C is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality laboratory applications.