Learn More
Hach Company Sension+ 9662 Nitrate Combination Ion Selective Electrode (ISE)
Catalog No. 501998027 Shop All Hach Company Products
Encompass
Sension+ 9662C is a combination nitrate ion selective electrode (ISE) with an epoxy body, non-refillable, double-junction reference, and non-replaceable, solid-state PVC (polymer) membrane selective to nitrate ions in solution. The 9662C has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM340 GLP pH/ISE Laboratory Meter or the Sension+ MM374 Laboratory Multimeter. The 9662C is ideal for measuring nitrate concentrations in drinking water, wastewater and general water quality laboratory applications.
Catalog No. 50-199-8027 Supplier Hach Company Supplier No. LZW9662C.97.002
Sension+ 9662C is a combination nitrate ion selective electrode (ISE) with an epoxy body, non-refillable, double-junction reference, and non-replaceable, solid-state PVC (polymer) membrane selective to nitrate ions in solution. The 9662C has a fixed 1 meter cable and BNC connector for laboratory use with the Hach Sension+ MM340 GLP pH/ISE Laboratory Meter or the Sension+ MM374 Laboratory Multimeter. The 9662C is ideal for measuring nitrate concentrations in drinking water, wastewater and general water quality laboratory applications.