Learn More
Genesee Scientific Corporation 96CELL PCR PLATE NON-SKRTD 25/
SDP
Catalog No. NC2396115
Encompass
Ultra Thin Wall, Natural25 Plates/UnitBrand: Olympus Plastics96-Well Non-Skirted, Flat-Top, 0.2ml wellsUltra-thin wall for optimum heat transfer and higher yieldsUltra clear, easy to visualize well contentsPlates can be sealed with strip caps, sealing films or silicone sealing matsBold, black alphanumeric indexingCertified free of detectable RNase, DNase and human DNAMade of 100% virgin polypropylene
Catalog No. NC2396115 Supplier Genesee Scientific Corporation Supplier No. 24300
Ultra Thin Wall, Natural25 Plates/UnitBrand: Olympus Plastics96-Well Non-Skirted, Flat-Top, 0.2ml wellsUltra-thin wall for optimum heat transfer and higher yieldsUltra clear, easy to visualize well contentsPlates can be sealed with strip caps, sealing films or silicone sealing matsBold, black alphanumeric indexingCertified free of detectable RNase, DNase and human DNAMade of 100% virgin polypropylene