Learn More
USA Scientific Inc 96-place tube rack, yellow
Catalog No. 502744749
Encompass
Polypropylene 96-place tube BestRack, yellow, 8 7/16 in L x 4 5/8 in W x 2 in H, 5/pack
Catalog No. 50-274-4749 Supplier USA Scientific Inc Supplier No. 23965006
Polypropylene 96-place tube BestRack, yellow, 8 7/16 in L x 4 5/8 in W x 2 in H, 5/pack