Learn More
Analytical Sales & Services 96POSITION DISCOVERY LED ARRAY
Supplier:  Analytical Sales & Services LUM296LAG1
Encompass
Discovery 1Lumidox® II, 96-Well Discovery LED Array (3 UVs)• Wavelengths: UV375nm, UV385nm, UV395nm• Each wavelength features 4 columns of 8 LEDs• Lens Mat• Used for screeningQty/Pkg: EachANALYTICAL SALES & SERVICES mfg#LUM296LAG1
Catalog No. NC2054489