Learn More
ALPAQUA ENGINEERING LLC 96S Super Magnet Plate
Catalog No. NC0284961
Encompass
The 96S magnet plate from Alpaqua contains stronger magnets for large volume separations of up to 2 ml, or for faster separations of smaller volumes. The 96S is also ideal for separation of more viscous samples, such as blood or serum.
Catalog No. NC0284961 Supplier ALPAQUA ENGINEERING LLC Supplier No. A001322
The 96S magnet plate from Alpaqua contains stronger magnets for large volume separations of up to 2 ml, or for faster separations of smaller volumes. The 96S is also ideal for separation of more viscous samples, such as blood or serum.