Learn More
Arcticwhite LLC 96 Square Deep Well, 1 ML, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS Base and Footprint, DNase-free, RNase-free, 50 Deep Well Plates per Case
SDP
Catalog No. NC0140033
Encompass
96 Square Deep Well, 1 ML, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS Base and Footprint, DNase-free, RNase-free, 50 Deep Well Plates per Case. This deep well microplate stores and processes samples for molecular biology and genomics. Format - 96-well, ANSI/SLAS base and footprint. Well shape - Square. Bottom - Conical. Well volume - 1 mL. Material - Polypropylene. Certifications - DNase-free, RNase-free. Supplied by ArcticWhite as 50 deep well plates per case.
Catalog No. NC0140033 Supplier Arcticwhite LLC Supplier No. AWLS219008
96 Square Deep Well, 1 ML, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS Base and Footprint, DNase-free, RNase-free, 50 Deep Well Plates per Case. This deep well microplate stores and processes samples for molecular biology and genomics. Format - 96-well, ANSI/SLAS base and footprint. Well shape - Square. Bottom - Conical. Well volume - 1 mL. Material - Polypropylene. Certifications - DNase-free, RNase-free. Supplied by ArcticWhite as 50 deep well plates per case.