Learn More
Arcticwhite LLC 96 Square Deep Well, 2.1 ML/Well, Polypropylene, Round Bottom, SBS Standard Footprint, Medical-grade Polypropylene, 24 Deep Well Plates per Case
SDP
Catalog No. NC9078451
Encompass
96 Square Deep Well, 2.1 ML/Well, Polypropylene, Round Bottom, SBS Standard Footprint, Medical-grade Polypropylene,-86 Deg C, This 96-well deep well microplate holds 2.1 mL per well. It suits automated liquid handling, nucleic acid purification, bacterial or mammalian cell culture, and sample storage. Certifications - SBS standard. Supplied by ArcticWhite as 24 deep well plates per case.
Catalog No. NC9078451 Supplier Arcticwhite LLC Supplier No. AWLST11010
96 Square Deep Well, 2.1 ML/Well, Polypropylene, Round Bottom, SBS Standard Footprint, Medical-grade Polypropylene,-86 Deg C, This 96-well deep well microplate holds 2.1 mL per well. It suits automated liquid handling, nucleic acid purification, bacterial or mammalian cell culture, and sample storage. Certifications - SBS standard. Supplied by ArcticWhite as 24 deep well plates per case.