Learn More
Stellar Scientific 96 Well Polystyrene Tissue Culture Treated Plate with V-Bottom, With Lid, RNase and DNase Free, Individually Wrapped, STERILE, 100/CS
SDP
Catalog No. NC2858121
Encompass
These 96-well tissue culture treated plates are made from high optic transmission grade polystyreneEach well can holds between 0.1-0.2mL
Catalog No. NC2858121 Supplier Stellar Scientific Supplier No. NST701201
These 96-well tissue culture treated plates are made from high optic transmission grade polystyreneEach well can holds between 0.1-0.2mL