Learn More
Chemglass Life Sciences 96 Well Evaporator/Concentrator Manifold, Fixed Needles
SDP
Catalog No. 507513684
Encompass_Preferred
96 Well Evaporator/Concentrator Manifold, Fixed Needles. Manifold is a low cost solution for the concentration of product in 96, 48 or 24 well blocks. Delivers equal amounts of gas to each well to gently speed solvent evaporation. Needles are constructed of stainless steel. 96 well unit has the needed pressed into the block. Manifold is constructed of polypropylene and the base is coated aluminum. Equipped with an adjustable flow meter for controlling gas flow into the manifold. Inlet is a 1/8” barbed fitting.Note: Blocks are not included and must be purchased separately.
Catalog No. 50-751-3684 Supplier Chemglass Life Sciences Supplier No. OP502001
96 Well Evaporator/Concentrator Manifold, Fixed Needles. Manifold is a low cost solution for the concentration of product in 96, 48 or 24 well blocks. Delivers equal amounts of gas to each well to gently speed solvent evaporation. Needles are constructed of stainless steel. 96 well unit has the needed pressed into the block. Manifold is constructed of polypropylene and the base is coated aluminum. Equipped with an adjustable flow meter for controlling gas flow into the manifold. Inlet is a 1/8” barbed fitting.Note: Blocks are not included and must be purchased separately.