Learn More
Agilent Technologies 96W NON-SKIRTED QPCR PLT 25/PK
Catalog No. NC9609903
Encompass
96-Well Non-Skirted Polypropylene PCR Plate for use with Mx3000P/Mx3005P QPCR Systems. Compatible with our Mx3000P/Mx3005P Optical Strip Caps, catalog number 401425.
Catalog No. NC9609903 Supplier Agilent Technologies Supplier No. 401333
96-Well Non-Skirted Polypropylene PCR Plate for use with Mx3000P/Mx3005P QPCR Systems. Compatible with our Mx3000P/Mx3005P Optical Strip Caps, catalog number 401425.