Learn More
NEST SCIENTIFIC INC 96 Well Cell Culture Plate, Flat, Non-Treated, sterile 100/cs
Catalog No. NC2945547
Encompass_Preferred
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are untreated to support suspended lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC2945547 Supplier NEST SCIENTIFIC INC Supplier No. 701011
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are untreated to support suspended lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.