Learn More
Thomas Scientific 96 Deep Well MASTERBLOCK Storage Plate, PP, 2mL/well, Square Wells, Conical Bottom, Natural, Bulk, 50/case
SDP
Catalog No. 50320867
Encompass_Preferred
Microplates are manufactured from virgin polypropylene resin, and feature uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics. Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance (–196 to 121°C/–321 to 250°F), and are resistant to many standard laboratory chemicals. These properties provide optimal conditions for laboratory specimen and compound storage. Ideal for cultivating bacteria or yeasts. Microplates feature alphanumerically coded wells. Conical well tops aid in complete sample recovery. Plates are autoclavable and supplied without lids. Microplates are RNase- and DNase-free, and nonpyrogenic. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
This item is not returnable. View return policy
Catalog No. 50-320-867 Supplier Thomas Scientific Supplier No. 1183A69
Microplates are manufactured from virgin polypropylene resin, and feature uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics. Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance (–196 to 121°C/–321 to 250°F), and are resistant to many standard laboratory chemicals. These properties provide optimal conditions for laboratory specimen and compound storage. Ideal for cultivating bacteria or yeasts. Microplates feature alphanumerically coded wells. Conical well tops aid in complete sample recovery. Plates are autoclavable and supplied without lids. Microplates are RNase- and DNase-free, and nonpyrogenic. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.