Learn More
Thomas Scientific 96W Plate, PS, Sterile, Round (U) Bottom, Clear, 100/case
SDP
Catalog No. 50320895
Encompass_Preferred
Free of detectable DNase, RNase, human DNA. Non-pyrogenic. Manufactured under DIN ISO 9001 guidelines. Can be traced all the way back to production through a defined LOT number system. Ideally suited for agglutination tests. No sharp corners, which means that they can be pipetted easily and cleanly. Suitable for +/- analyses. Solid bottom.
Catalog No. 50-320-895 Supplier Thomas Scientific Supplier No. 1184J43
Free of detectable DNase, RNase, human DNA. Non-pyrogenic. Manufactured under DIN ISO 9001 guidelines. Can be traced all the way back to production through a defined LOT number system. Ideally suited for agglutination tests. No sharp corners, which means that they can be pipetted easily and cleanly. Suitable for +/- analyses. Solid bottom.