Learn More
Electron Microscopy Sciences Micro Centrifuge Tubes 96-Well Reversible Rack, Assorted Colors
SDP
Catalog No. NC1625469
Encompass_Preferred
Polypropylene rack has 96 wells to accommodate 0.5 ml micro centrifuge tubes on one side, or 96 wells for 1.5 to 2.0 ml micro centrifuge tubes on the other side. Arranged in standard 8 x 12 array. Handy lid snaps into place in the tabs on each end of rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the opposite side of the rack making it easier to replace the 1.5 to 2.0 ml tubes from those wells.
Rack measures 9.75 x 4.75 x 1.875".
Color: Assorted Colors
Catalog No. NC1625469 Supplier Electron Microscopy Sciences Supplier No. 61602A
Polypropylene rack has 96 wells to accommodate 0.5 ml micro centrifuge tubes on one side, or 96 wells for 1.5 to 2.0 ml micro centrifuge tubes on the other side. Arranged in standard 8 x 12 array. Handy lid snaps into place in the tabs on each end of rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the opposite side of the rack making it easier to replace the 1.5 to 2.0 ml tubes from those wells.
Rack measures 9.75 x 4.75 x 1.875".
Color: Assorted Colors