Learn More
Arcticwhite LLC ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, Pre-Slit, 5 Plate Seals per Pack
SDP
Catalog No. NC9947861
Encompass
ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, Pre-Slit, 5 Plate Seals per Pack. Sealing mat for 96-well microplates used in PCR, liquid handling, and high-throughput screening. Supplied by ArcticWhite as 5 plate seals per pack.
Catalog No. NC9947861 Supplier Arcticwhite LLC Supplier No. AWSM1002RBX
ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, Pre-Slit, 5 Plate Seals per Pack. Sealing mat for 96-well microplates used in PCR, liquid handling, and high-throughput screening. Supplied by ArcticWhite as 5 plate seals per pack.