Learn More
Arcticwhite LLC ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, 5 Plate Seals per Pack
SDP
Catalog No. NC9743907
Encompass
ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, 5 Plate Seals per Pack. This sealing mat is molded from silicone. It fits 96-well plates with round wells. It has a PTFE coating. This variant is white and has a round bottom. ArctiSeal sealing mats and strips seal 96-well microplates for PCR, liquid handling, and high-throughput screening. The range fits round well and square well formats, bottom geometries, and laboratory automation systems. Supplied by ArcticWhite as 5 plate seals per pack.
Catalog No. NC9743907 Supplier Arcticwhite LLC Supplier No. AWSM1002RB
ArctiSeal 96 Well Round, White Silicone, PTFE Coating, Round Bottom, 5 Plate Seals per Pack. This sealing mat is molded from silicone. It fits 96-well plates with round wells. It has a PTFE coating. This variant is white and has a round bottom. ArctiSeal sealing mats and strips seal 96-well microplates for PCR, liquid handling, and high-throughput screening. The range fits round well and square well formats, bottom geometries, and laboratory automation systems. Supplied by ArcticWhite as 5 plate seals per pack.