Learn More
Arcticwhite LLC 96 Round Well, 0.5 ML/Well, Polypropylene, Round Bottom, 96 Wells, DNase/RNase Free, 50 Microplates per Case
SDP
Catalog No. NC9015318
Encompass
96 Round Well, 0.5 ML/Well, Polypropylene, Round Bottom, 96 Wells, DNase/RNase Free, 50 Microplates per Case. This 96-well polypropylene microplate stores and processes samples in molecular biology workflows. Height - 14.3 mm. Certifications - DNase/RNase free. Supplied by Arctic White as 50 microplates per case.
Catalog No. NC9015318 Supplier Arcticwhite LLC Supplier No. AWLSX10001
96 Round Well, 0.5 ML/Well, Polypropylene, Round Bottom, 96 Wells, DNase/RNase Free, 50 Microplates per Case. This 96-well polypropylene microplate stores and processes samples in molecular biology workflows. Height - 14.3 mm. Certifications - DNase/RNase free. Supplied by Arctic White as 50 microplates per case.