Learn More
Research Products International Corp RPI 96 Well Assay Plate, Non-Sterile, Flat Bottom without Lid, 10 per Bag, 100 per Case
SDP
Catalog No. 50476447
Encompass
Assay plates are designed for all laboratory applications requiring a 96-well format. Plates feature standardized dimensions and are compatible with all automated plate handling systems. Recommended working volume is 40-280ul per well. Ridges on plates provide stable stacking on a bench top or incubator.
Catalog No. 50-476-447 Supplier Research Products International Corp Supplier No. 141383
Add to Cart
Add to Cart
Assay plates are designed for all laboratory applications requiring a 96-well format. Plates feature standardized dimensions and are compatible with all automated plate handling systems. Recommended working volume is 40-280ul per well. Ridges on plates provide stable stacking on a bench top or incubator.