Learn More
Research Products International Corp RPI 96 Well Assay Plate, Sterile, Flat Bottom with Lid, Individually Wrapped, 100 per Case
SDP
Supplier:  Research Products International Corp 141380
Encompass
Assay plates are designed for all laboratory applications requiring a 96-well format. Plates feature standardized dimensions and are compatible with all automated plate handling systems. Recommended working volume is 40-280ul per well. Ridges on plates provide stable stacking on a bench top or incubator.
Catalog No. 50-476-445