Learn More
Alkali Scientific 96 Well Cell Culture Non-Treated Plate, White, Sterile, Individually Wrapped, 50/cs
SDP
Catalog No. 502335934
Encompass_Preferred
▪ HTS (High-Throughput Screening) compatible ▪ Suitable for fluorescence assay, luminescence assay ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Homogeneous cell culture ▪ Flat bottom ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant
Catalog No. 50-233-5934 Supplier Alkali Scientific Supplier No. TPW96
▪ HTS (High-Throughput Screening) compatible ▪ Suitable for fluorescence assay, luminescence assay ▪ Designed to prevent cross contamination ▪ Effective gas exchange lid inner design ▪ Alphanumeric labeling ▪ Homogeneous cell culture ▪ Flat bottom ▪ USP guideline compliant (USP class Ⅵ tested) ▪ ANSI guideline compliant