Learn More
Cole-Parmer Essentials 96-well DispoDialyzer, membrane MWCO 10,000 Daltons
Catalog No. NC1697988 Shop All Cole-Parmer Products
Encompass_Preferred
These 96-well dialyzers let you simultaneously prepare 96 samples. Use for applications such as salt removal, buffer exchange, parallel sample prep after fraction collection, detergent removal, HPLC, and more. Each well contains an individual, high-quality, regenerated cellulose membrane that eliminates sample contamination and leakage. Membranes are free from sulfur and heavy metal contamination. Wells are sealed with 8-cap strips (included). To use the dialyzer, simply place the sample into each well and assemble the strip caps. Float the entire plate in dialysis buffer. Then, retrieve the purified samples for downstream applications. What¿s included: twelve 8-cap strips to seal wells.
Catalog No. NC1697988 Supplier Cole-Parmer Supplier No. 0289286
These 96-well dialyzers let you simultaneously prepare 96 samples. Use for applications such as salt removal, buffer exchange, parallel sample prep after fraction collection, detergent removal, HPLC, and more. Each well contains an individual, high-quality, regenerated cellulose membrane that eliminates sample contamination and leakage. Membranes are free from sulfur and heavy metal contamination. Wells are sealed with 8-cap strips (included). To use the dialyzer, simply place the sample into each well and assemble the strip caps. Float the entire plate in dialysis buffer. Then, retrieve the purified samples for downstream applications. What¿s included: twelve 8-cap strips to seal wells.